missing translation for 'onlineSavingsMsg'
Learn More

Pirenzepine dihydrochloride, 99%, Thermo Scientific™

A M1 muscarinic receptor antagonist

Brand:  Thermo Scientific Alfa Aesar J62252.03

Quantity 1g

 View more versions of this product

Product Code. 15424659

  • ONLINE EXCLUSIVE PRICE
    £138.00
    £135.01 / 1g
    Save £2.99
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

29868-97-1
424.326
FFNMBRCFFADNAO-UHFFFAOYSA-N
71405
11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
C19H23Cl2N5O2
MFCD00055214
pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
CHEBI:32014
CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl

Specifications

29868-97-1
MFCD00055214
Hygroscopic
pirenzepine dihydrochloride, pirenzepine hydrochloride, tabe, bisvanil, leblon, maghen, pirenzepine hcl, pirenzepine 2hcl, ls 519 dihydrochloride, unii-10ym403fls
CN1CCN(CC1)CC(=O)N2C3=CC=CC=C3C(=O)NC4=C2N=CC=C4.Cl.Cl
424.326
CHEBI:32014
99%
C19H23Cl2N5O2
1g
14,7491
FFNMBRCFFADNAO-UHFFFAOYSA-N
11-[2-(4-methylpiperazin-1-yl)acetyl]-5H-pyrido[2,3-b][1,4]benzodiazepin-6-one;dihydrochloride
71405
424.32
Pirenzepine dihydrochloride
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.