missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pirarubicin Hydrochloride, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Description
Pirarubicin Hydrochloride is an anthracycline antibiotics, acts as a topoisomerase II inhibitor, and is a widely used for treatment of various cancers, in particular, solid tumors.
Specifications
Specifications
| Chemical Name or Material | Pirarubicin Hydrochloride |
| Physical Form | Solid |
| CAS | 95343-20-7 |
| Color | Red |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C32H38ClNO12 |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | THP Hydrochloride |
| Solubility Information | DMSO : 20.83 mg/mL (31.37 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction