missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10263-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the cupin superfamily. The encoded protein is an Fe(II)-containing nuclear protein expressed in all tissues of the body and concentrated within dot-like subnuclear structures. Interactions with nuclear factor I/CCAAT box transcription factor as well as B cell lymphoma 3-encoded oncoprotein suggest the encoded protein may act as a transcriptional cofactor and be involved in the regulation of DNA transcription and replication. Alternatively spliced transcript variants have been described.Specifications
| PIR | |
| Polyclonal | |
| Unconjugated | |
| PIR | |
| PIR, Pirin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 363465, 69656, 8544 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.14 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O00625, Q5M827, Q9D711 | |
| Pir | |
| PIR Fusion Protein Ag0330 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction