Learn More
Invitrogen™ PIR-A/B Monoclonal Antibody (10-1-PIR), APC, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
Brand: Invitrogen™ 17-3101-82
Description
Description: This 10-1-PIR monoclonal antibody reacts with mouse paired Ig-like receptors of activating (PIR)-A and -B. These cell surface glycoproteins, which contain six extracellular Ig-like domains with distinct transmembrane and cytoplasmic regions, are expressed together on B lymphocytes, dendritic cells, macrophages, granulocytes, platelets, and mast cells. However, expression of PIR-A/B has not been observed on T cells, NK cells, and erythrocytes. The PIR-A receptor interacts with signaling molecules containing immunoreceptor tyrosine-based activation motifs (ITAMs) which lead to its activating function. In contrast, the PIR-B receptor associates with proteins possessing immunoreceptor tyrosine-based inhibitory motifs (ITIMs); therefore, this receptor has been shown to have an inhibitory function. PIRs bind MHC class I to modulate cell signaling and homeostasis of the immune system. Moreover, PIR-B knockout mice have been shown to exhibit susceptibility to Salmonella infection. Crossblocking studies indicate that 10-1-PIR recognizes a different epitope from 6C1 (cat. no. 46-5978). Applications Reported: This 10-1-PIR antibody has been reported for use in flow cytometric analysis. Applications Tested: This 10-1-PIR antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test.
Paired Ig-like receptors of activating (PIR)-A and -B are expressed on B lymphocytes, macrophages, dendritic cells, platelets, granulocytes, and mast cells, but not on T cells, NK cells, and erythrocytes. These cell surface glycoproteins contain six extracellular Ig-like domains with distinct cytoplasmic and transmembrane regions. The PIR-A receptor activates through signaling molecules containing immunoreceptor tyrosine-based activation motifs. In contrast, the PIR-B receptor inhibits through proteins possessing immunoreceptor tyrosine-based inhibitory motifs. PIRs bind MHC class I to modulate immune system homeostasis and cell signaling.
Specifications
| PIR-A/B | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P97484 | |
| Pira1, Pirb | |
| Affinity chromatography | |
| RUO | |
| 18722, 18733 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| 10-1-PIR | |
| APC | |
| Pirb | |
| 6M21; CD85k; cell-surface glycoprotein p91; Gp49b; Gp91; leukocyte immunoglobulin-like receptor 3; leukocyte immunoglobulin-like receptor subfamily B member 3; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; Lilrb3; Lilrb4; LIR-3; Ly89; paired Ig-like receptor B; paired immunoglobulin-like receptor; paired immunoglobulin-like receptor B; paired-Ig-like receptor A1; Pir; Pira1; PIR-A1; Pirb; PIR-B | |
| Rat | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.