missing translation for 'onlineSavingsMsg'
Learn More

Pipsyl chloride, 97%

Product Code. 10032710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10032710 25 g Glass Bottle 25g
10184880 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10032710 Supplier Thermo Scientific Acros Supplier No. 207290250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98-61-3 | C6H4ClIO2S | 302.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-61-3
Molecular Formula C6H4ClIO2S
Molecular Weight (g/mol) 302.51
MDL Number MFCD00007441
InChI Key POXFXTSTVWDWIR-UHFFFAOYSA-N
Synonym pipsyl chloride, 4-iodobenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 4-iodo, p-iodobenzenesulfonyl chloride, 4-iodobenzenesulphonyl chloride, 4-iodobenzenesulfonylchloride, 4-iodophenylsulfonyl chloride, iodobenzene-p-sulfonyl chloride, unii-mvl274ezog, mvl274ezog
PubChem CID 7399
IUPAC Name 4-iodobenzenesulfonyl chloride
SMILES C1=CC(=CC=C1S(=O)(=O)Cl)I

Specifications

Melting Point 80°C to 84°C
Color Beige to White or Orange
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula IC6H4SO2Cl
Quantity 25 g
Beilstein 11, 65
Merck Index 15, 7599
Formula Weight 302.51
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Pipsyl chloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Pipsyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.