missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PIPOX Polyclonal Antibody

Description
The antibody detects endogenous levels of total PIPOX protein.
Metabolizes sarcosine, L-pipecolic acid and L-proline.
Specifications
Specifications
| Antigen | PIPOX |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Pipox |
| Gene Accession No. | Q9D826, Q9P0Z9 |
| Gene Alias | L-pipecolate oxidase; L-pipecolic acid oxidase; LPIPOX; Peroxisomal sarcosine oxidase; pipecolic acid and sarcosine oxidase; pipecolic acid oxidase; PIPOX; Pso |
| Gene Symbols | Pipox |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?