missing translation for 'onlineSavingsMsg'
Learn More

Pipobroman, TRC

Product Code. 30415621
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30415621 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30415621 Supplier Toronto Research Chemicals Supplier No. TRCP4906001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-91-1
Molecular Formula C10H16Br2N2O2
Molecular Weight (g/mol) 356.05
Synonym 1,1'-(1,4-Piperazinediyl)bis[3-bromo-1-propanone], 1,4-bis(3-Bromo-1-oxopropyl)piperazine, 1,4-bis(3-Bromopropionyl)piperazine, A 1803, Amedel, N,N'-Bis(3-bromopropionyl)piperazine, NSC-25154, Vercyte
IUPAC Name 3-bromo-1-[4-(3-bromopropanoyl)piperazin-1-yl]propan-1-one
SMILES BrCCC(=O)N1CCN(CC1)C(=O)CCBr

Specifications

Quantity 1 g
InChI Formula InChI=1S/C10H16Br2N2O2/c11-3-1-9(15)13-5-7-14(8-6-13)10(16)2-4-12/h1-8H2
Formula Weight 353.9579
Chemical Name or Material Pipobroman
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.