missing translation for 'onlineSavingsMsg'
Learn More

Pipobroman, MedChemExpress

Product Code. 16477096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16477096 Supplier Medchem Express Supplier No. HY16398

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pipobroman is a bromide derivative of piperazine and acts as an alkylating agent. Pipobroman plays its role by inhibiting DNA and RNA polymerase or by reducing pyrimidine nucleotide incorporation into DNA. Pipobroman can be used for the cancer research, including polycythemia vera, myeloproliferative neoplasm, and AML et.al.

Pipobroman is a bromide derivative of piperazine and acts as an alkylating agent. Pipobroman plays its role by inhibiting DNA and RNA polymerase or by reducing pyrimidine nucleotide incorporation into DNA. Pipobroman can be used for the cancer research, including polycythemia vera, myeloproliferative neoplasm, and AML et.al.

Specifications

Chemical Name or Material Pipobroman
Physical Form Solid
CAS 54-91-1
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C10H16Br2N2O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 36 mg/mL (101.11 mM)
SMILES O=C(N1CCN(C(CCBr)=O)CC1)CCBr
Molecular Weight (g/mol) 356.05
Formula Weight 356.05
Percent Purity 95.06%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.