missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Microloader™ Pipette Tips

Product Code. 10289651
Encompass
Change view
Click to view available options
Volume (Metric):
0.5 to 20 μL
Unit Size:
Pack of 192
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10289651 0.5 to 20 μL Pack of 192
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10289651 Supplier Eppendorf™ Supplier No. 5242956003

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Use these extremely long, fine and flexible tips for filling of microcapillaries. Eppendorf ™ Microloader™ Pipette Tips are ideal for micro-injection and where additional reach is needed.

  • For filling microinjection capillaries
  • 0.5μL to 20μL
  • Racked

Specifications

Volume (Metric) 0.5 to 20 μL
For Use With (Application) For filling microinjection capillaries
Format Rack
For Use With (Equipment) Eppendorf Microinjection Capillaries 0.5 μL to 20 μL
Product Line Microloader
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.