Learn More
BRAND™ Filter Tips, 5 - 100 μL, PP, Colorless, Filter PE, BIO-CERT™ Liquid Handling Quality
Product Code. 15144957 Shop All Brand Products
Change view
Click to view available options
Volume (Metric):
5 to 100 μL
Unit Size:
Pack of 960
Product Code. Volume (Metric) Unit Size
15144957 5 to 100 μL Pack of 960
1 options
Product Code. 15144957 Supplier BRAND™ Supplier No. 732610
  • ONLINE EXCLUSIVE PRICE
    £79.75
    £78.35 / Pack of 960
    Save £1.40
    1% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Bulk Quote Request
missing translation for 'validMainframeMsg'
Cleanroom quality for contamination-free analyses. Perfect fit for the Transferpette™ micropipette and many other pipettes. The tip system for all requirements.
Pipette tips and filter tips from BRAND offer reliable quality for precise and demanding analyses. The raw materials are free from DiHEMDA and Oleamid. Tips are manufactured at BRAND under monitored cleanroom conditions and are automatically packaged. No parting agents or demolding aids are used. BRAND offers pipette tips for volumes from 0.1 μl to 10 ml and filter tips for volumes from 0.1 μl to 1250 μl. They are tested for BRAND pipettes and many pipette from other manufacturers.
  • BRAND pipette tips and filter tips as well as the corresponding TipBoxes are autoclavable at 121 °C (2 bar) acc. DIN EN 285.
  • Sterile tips are manufactured under certified BIO-CERT™ quality. They are sterilized with beta radiation according to ISO 11 137 and the AAMI Guidelines.
  • Non-self-sealing filter tips from BRAND have a PE filter that is free from chemical additives. Permeability is controlled by the distinct combination of pore size and filter length, so that no aerosols can reach the pipette shaft.
  • BIO-CERT™ Liquid Handling Quality: Free of human DNA, RNase, DNase, Pyrogens, PCR-Inhibitors and ATP
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 5 to 100 μL
Sterility Non-sterile
Certifications/Compliance DIN EN 285
Packaging Bag
Quantity 960/PK
Color Code Yellow
Material Polypropylene