missing translation for 'onlineSavingsMsg'
Learn More

VistaLab™ Technologies Pipetter Nozzles

Product Code. p-4924578
Click to view available options
Volume (Metric):
0.5 to 20 μL
100 to 200 μL
2 μL
2 to 20 μL
5 mL
Unit Size:
Pack of 10
Pack of 2
Pack of 4
Pack of 5
This item is not returnable. View return policy

Product Code. 11976022

Brand: VistaLab™ Technologies 90572010

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Contains an internal aerosol/liquid barrier filter to prevent liquid from being aspirated into pipetter

  • Available in range of sizes

Specifications

For Use With Pipettes
Material Plastic
Sterility Non-sterile
Volume (Metric) 100 to 200 μL
For Use With (Equipment) 2 to 125 μL Electronic Single Channel Model, 5 to 250 μL and 20 to 200 μL Adjustable Model, 21 to 200 μL Fixed Models
Includes Internal aerosol/liquid barrier filter
Quantity 10/Box
Length (English) 5 in.
Width (English) 3 in.
Height (English) 1 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.