missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Rubber Pipet Suction Bulb
Click to view available options
Color:
Green
Unit Size:
Each
Description
- For liquid handling
Specifications
Specifications
| Color | Green |
| Material | Rubber |
| Type | Suction Bulb |
| Volume (Metric) | 1 mL |
| For Use With (Application) | For fast and easy pipetting |
| Quantity | 1 Each |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction