missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pipette microcaps disposable borosilicate glass 116mm length 100μL
Brand: Cole Parmer 21170N
Code : 02
Additional Details : Weight : 0.20000kg
Brand: Cole Parmer 21170N
Code : 02
Additional Details : Weight : 0.20000kg