Learn More
VITLAB™ Pipette Fillers
Product Code. p-8006524
Click to view available options
For Use With (Equipment):
2 mL Pipettes
Up to 10 mL Pipettes
Unit Size:
Each
Pack of 10
Product Code. 10489382
Brand: VITLAB™ 104099
Measure the volume of liquids using Vitlab™ Pipette Filters. These filters draw liquid into the pipette.
  • Slow rotation of the actuator-wheels draws liquid into the pipette
  • Pressing air bleed valve automatically empties the pipette without returning the piston
  • Green, Blue and Red available
Specifications
For Use With (Equipment) Up to 10 mL Pipettes