Learn More
VITLAB™ Pipette Fillers
Click to view available options
For Use With (Equipment):
2 mL Pipettes
Up to 10 mL Pipettes
Unit Size:
Each
Pack of 10
Description
- Slow rotation of the actuator-wheels draws liquid into the pipette
- Pressing air bleed valve automatically empties the pipette without returning the piston
- Green, Blue and Red available
Specifications
Specifications
| For Use With (Equipment) | 2 mL Pipettes |