Description
- Accessory item
- Kit consists of a moisture trap and all required adapters
Specifications
Specifications
| For Use With (Equipment) | Lab Balance with 0.01 mg readability |
| For Use With (Application) | Weighing |
| Includes | Moisture trap and adapters |
| Type | Pipette Calibration Kit |