Learn More
Cole Parmer Pipette Bulb
Cole Parmer Pipette Bulb is sturdy, re-usable and easy-to-use.
Brand: Cole Parmer 24820-60
Code : A0
Additional Details : Weight : 0.80000kg
Description
Cole Parmer Pipette Bulb is sturdy, re-usable and easy-to-use.
- Made from natural rubber and silicone
- Can be used with Pasteur, serological, volumetric and other standard pipettes
- Silicone seal grips the glass firmly and protects it
Specifications
Rubber, Silicone | |
Non-sterile | |
25 mL |