Learn More
Gilson™ PIPETMAN™ Comfort Handle
The ergonomically designed PIPETMAN™ Comfort Handle attaches to any PIPETMAN Classic, Neo, or G pipette to provide improved comfort and reduced pipetting fatigue.
Brand: Gilson™ F161487
Additional Details : Weight : 0.25000kg
Description
- Large, more comfortable finger hook and ergonomically designed handle
- Fits better in your hand
- Offers better grip to support your daily pipetting activity and aids in reducing the risk of repetitive strain injuries
- Contains an identification zone that allows you to mark the pipette using PIPETMAN™ Comfort Handle Labels
- Easily attached to the pipette with no tools required
- Labels are easily removed and the handle is autoclavable
- White in color
- Provided in packs of 50
Specifications
Autoclavable | |
Ergonomic pipetting | |
PIPETMAN™ pipetting system | |
50/Pk. |