Learn More
Mettler Toledo™ Pipet-Lite™ XLS Model 8-Channel Variable Volume Manual Pipette
Product Code. 12721032
Encompass
Change view
Click to view available options
Volume (Metric):
300 μL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
12721032 300 μL Each
1 options
Product Code. 12721032 Supplier Mettler Toledo™ Supplier No. 17011843
Only null left
Add to basket
Add to basket
Specifications
Model XLS+
Number of Channels 8
Product Line Pipet-Lite™ XLS
Type Variable Volume Manual Pipette
Volume (Metric) 300 μL
For Use With (Application) Genomic, Proteomic, Tissue Culture and Cell Culture applications
Quantity 1 Pack