missing translation for 'onlineSavingsMsg'
Learn More

PIPES, Sesquisodium Sodium Salt, Fisher BioReagents

Product Code. p-27672
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10488723

Brand: Fisher BioReagents BP304100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Commonly used buffering agent

This biological buffer has a usable pH range of 6.1 to 7.5.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100037-69-2
Molecular Formula C16H33N4Na3O12S4
Molecular Weight (g/mol) 670.665
InChI Key FPDACHMSOMILQI-UHFFFAOYSA-K
Synonym 1, 4-Piperazinediethanesulfonic acid sesquisodium salt, Piperazine-1, 4-bis(2-ethanesulfonic acid) sesquisodium salt
PubChem CID 3086462
IUPAC Name trisodium;2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonate;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)[O-].C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 300°C
Color White
pH 6.5 to 7.1
Physical Form Powder or Solid
Quantity 100 g
DNase DNase free
Assay Percent Range ≥95 %
Identification Pass Test
Packaging Poly Bottle
Protease Protease free
Formula Weight 670.68
Percent Purity ≥95%
Grade DNase- and RNase-Free
Chemical Name or Material PIPES, Sesquisodium Salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.