missing translation for 'onlineSavingsMsg'
Learn More

Melford PIPES [Piperazine-N-N'-bis(2-ethanesulfonic acid)]

Product Code. 30018694
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
500 g
1 kg
Unit Size:
100g
10g
1kg
250g
500g
50g
This item is not returnable. View return policy

Product Code. 30018694

Brand: Melford P40140100.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.

Heavy metals: <10 ppm

pka (25°C): 6.8

The working pH range in aqueous solution is 6.5-7.5

Specifications

Chemical Name or Material PIPES
CAS 5625-37-6
Quantity 100 g
Molecular Formula C8H18N2O6S2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.