missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford PIPES [Piperazine-N-N'-bis(2-ethanesulfonic acid)]
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
500 g
1 kg
Unit Size:
100g
10g
1kg
250g
500g
50g
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Heavy metals: <10 ppm
pka (25°C): 6.8
The working pH range in aqueous solution is 6.5-7.5
Specifications
Specifications
| Chemical Name or Material | PIPES |
| CAS | 5625-37-6 |
| Quantity | 10 g |
| Molecular Formula | C8H18N2O6S2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction