missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PIPES, 99.4%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
500g
Specifications
Specifications
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 100 g |
| Assay Percent Range | 0.994 |
| Formula Weight | 302.4 |
| Chemical Name or Material | Pipes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction