missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals PIPES, 98.5+%, for biochemistry

Product Code. 10731414 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10731414 500 g Plastic bottle 500g
10711261 25 g Plastic bottle 25g
10062200 100 g Plastic bottle 100g
11956521 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10731414 Supplier Thermo Scientific Chemicals Supplier No. 172615000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 5625-37-6 | C8H18N2O6S2 | 302.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Biological buffer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5625-37-6
Molecular Formula C8H18N2O6S2
Molecular Weight (g/mol) 302.36
InChI Key IHPYMWDTONKSCO-UHFFFAOYSA-N
Synonym 1, 4-Piperazinebis(ethanesulfonic acid)
PubChem CID 79723
ChEBI CHEBI:44933
IUPAC Name 2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonic acid
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)O

Specifications

Absorbance (0.1M in 0.1M NaOH) (1 cm cell), (0.1M in 0.1M NaOH) (1 cm cell) at 0nm, 0.04 max. at 280nm, 0.05 max. at 260nm
Absorbance Conditions (0.1M in 0.1M NaOH) (1cm cell)
Color White
Physical Form Micro-crystalline Powder
Quantity 500 g
Assay Percent Range 98.5+%
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Packaging Plastic bottle
Formula Weight 302.35
Percent Purity ≥98.50%
Grade Biochemical
Chemical Name or Material PIPES
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.