missing translation for 'onlineSavingsMsg'
Learn More

Piperonylic Acid, TRC

Product Code. 30427968
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427968 10 g -
30425009 5 g -
30409162 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30427968 Supplier Toronto Research Chemicals Supplier No. TRCP49045010G
 Restricted Item
Estimated Shipment: 15-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-53-1
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
Synonym 1,3-Benzodioxole-5-carboxylic acid (9CI, ACI), Piperonylic acid (6CI, 7CI, 8CI), 1,3-Dioxaindane-5-carboxylic acid, 2H-1,3-Benzodioxole-5-carboxylic acid, 3,4-Dioxymethylenebenzoic acid, 3,4-Methylenedioxybenzenecarboxylic acid, 3,4-Methylenedioxybenzoic acid, 5-Benzodioxolecarboxylic acid, Benzo[d][1,3]dioxole-5-carboxylic acid, Heliotropic acid, NSC 10072, NSC 119055, Protocatechuic acid methylene ether
IUPAC Name 1,3-benzodioxole-5-carboxylic acid
SMILES OC(=O)c1ccc2OCOc2c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H6O4/c9-8(10)5-1-2-6-7(3-5)12-4-11-6/h1-3H,4H2,(H,9,10)
Formula Weight 166.0266
Chemical Name or Material Piperonylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.