missing translation for 'onlineSavingsMsg'
Learn More

Piperonylic acid hydrazide, 97%

Product Code. 11474620
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11474620 1 g 1g
11484620 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11474620 Supplier Thermo Scientific Alfa Aesar Supplier No. H50713.03
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22026-39-7 | C8H8N2O3 | 180.163 g/mol

Piperonylic acid hydrazide is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperonylic acid hydrazide is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22026-39-7
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.163
MDL Number MFCD00060505
InChI Key RAXBGBHBUFGWPG-UHFFFAOYSA-N
Synonym 3,4-methylenedioxybenzhydrazide, benzo d 1,3 dioxole-5-carbohydrazide, piperonylic acid hydrazide, 2h-1,3-benzodioxole-5-carbohydrazide, 3,4-methylenedioxybenzoic acid hydrazide, 3,4-methylenedioxy benzohydrazide, 1,3-benzodioxole-5-carboxylic acid, hydrazide, 3,4-methylenedioxy benzhydrazide, benzo 1,3 dioxole-5-carboxylic acid hydrazide, 2h-benzo d 1,3-dioxolane-5-carbohydrazide
PubChem CID 89158
IUPAC Name 1,3-benzodioxole-5-carbohydrazide
SMILES C1OC2=C(O1)C=C(C=C2)C(=O)NN

Specifications

Melting Point 176°C to 178°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 180.16
Percent Purity 97%
Chemical Name or Material Piperonylic acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.