missing translation for 'onlineSavingsMsg'
Learn More

Piperonylic acid, 99%

Product Code. 10172990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10172990 25 g Plastic bottle 25g
10636934 5 g Glass bottle 5g
10458100 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10172990 Supplier Thermo Scientific Acros Supplier No. 131370250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 94-53-1 | C8H6O4 | 166.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-53-1
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00005830
InChI Key VDVJGIYXDVPQLP-UHFFFAOYSA-N
Synonym piperonylic acid, heliotropic acid, 3,4-methylenedioxybenzoic acid, benzo d 1,3 dioxole-5-carboxylic acid, 2h-1,3-benzodioxole-5-carboxylic acid, benzo 1,3 dioxole-5-carboxylic acid, protocatechuic acid methylene ether, 5-benzodioxolecarboxylic acid, 3,4-dioxymethylenebenzoic acid, 3,4-methylene dioxybenzoic acid
PubChem CID 7196
IUPAC Name 1,3-benzodioxole-5-carboxylic acid
SMILES OC(=O)C1=CC=C2OCOC2=C1

Specifications

Melting Point 229°C to 235°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 19, 269
Merck Index 15, 7590
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in chloroform,cold alcohol and,ether
Formula Weight 166.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Piperonylic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Piperonylic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.,Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.,Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P312-Call a POISON CENTER/doctor/physician/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.