missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperonylamine, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | Piperonylamine |
| CAS | 2620-50-0 |
| Molecular Formula | C8H9NO2 |
| Quantity | 50 g |
| Synonym | 1,3-Benzodioxole-5-methanamine, (Benzo[d][1,3]dioxol-5-yl)methanamine, 1,3-Benzodioxol-5-ylmethanamine, 1,3-Benzodioxol-5-ylmethylamine, 1-(1,3-Benzodioxol-5-yl)methanamine, 1-(Aminomethyl)-3,4-(methylenedioxy)benzene, 3,4-(Methylenedioxy)benzylamine, 3,4-Methylenedioxyphenylmethylamine, 5-Aminomethyl-1,3-benzodioxole, NSC 75851, [(Benzo[d][1,3]dioxol-5-yl)methyl]amine, [(Benzodioxol-5-yl)methyl]amine |
| InChI Formula | InChI=1S/C8H9NO2/c9-4-6-1-2-7-8(3-6)11-5-10-7/h1-3H,4-5,9H2 |
| SMILES | NCc1ccc2OCOc2c1 |
| IUPAC Name | 1,3-benzodioxol-5-ylmethanamine |
| Molecular Weight (g/mol) | 151.16 |
| Formula Weight | 151.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction