missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl butoxide, MedChemExpress

Product Code. 16438676
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438676 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16438676 Supplier Medchem Express Supplier No. HYB1198
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Piperonyl butoxide is a semisynthetic derivative of safrole,used as a component of pesticide formulations. It is a synergist, despite having no pesticidal activity of its own, it enhances the potency of certain pesticides such as Carbamates, Pyrethrins, Pyrethroids, and Rotenone.

Piperonyl butoxide is a semisynthetic derivative of safrole,used as a component of pesticide formulations. It is a synergist, despite having no pesticidal activity of its own, it enhances the potency of certain pesticides such as Carbamates, Pyrethrins, Pyrethroids, and Rotenone.

Specifications

Chemical Name or Material Piperonyl butoxide
CAS 51-03-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H30O5
Quantity 100 mg
Synonym ENT-14250
Solubility Information DMSO : 100 mg/mL (295.47 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (295.47 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CCCC1=C(COCCOCCOCCCC)C=C2OCOC2=C1
Molecular Weight (g/mol) 338.44
Formula Weight 338.44
Percent Purity 97.0%
Grade Research
Physical Form Oil
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.