missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl Butoxide (~90%), TRC

Product Code. 30418203
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30418203 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30418203 Supplier Toronto Research Chemicals Supplier No. TRCP4902051G
 Restricted Item
Estimated Shipment: 27-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Piperonyl butoxide
CAS 51-03-6
Molecular Formula C19 H30 O5
Quantity 1 g
Synonym 1,3-Benzodioxole','5-[[2-(2-butoxyethoxy)ethoxy]methyl]-6-propyl-, Toluene','α-[2-(2-butoxyethoxy)ethoxy]-4,5-(methylenedioxy)-2-propyl- (8CI), 5-[[2-(2-Butoxyethoxy)ethoxy]methyl]-6-propyl-1,3-benzodioxole, 6-Propylpiperonyl butyl diethylene glycol ether, Butacide, Buticide, Butocide, Butoxide, Butoxide (synergist), Butyl carbitol 6-propylpiperonyl ether, ENT 14,250, Incite, NSC 8401, PBO, Piperonyl, Piperonyl butoxide, Zengxiaomi, α-[2-(2-Butoxyethoxy)ethoxy]-4,5-methylenedioxy-2-propyltoluene, α-[2-(2-n-Butoxyethoxy)-ethoxy]-4,5-methylenedioxy-2-propyltoluene
InChI Formula InChI=1S/C19H30O5/c1-3-5-7-20-8-9-21-10-11-22-14-17-13-19-18(23-15-24-19)12-16(17)6-4-2/h12-13H,3-11,14-15H2,1-2H3
SMILES CCCCOCCOCCOCc1cc2OCOc2cc1CCC
IUPAC Name 5-[2-(2-butoxyethoxy)ethoxymethyl]-6-propyl-1,3-benzodioxole
Molecular Weight (g/mol) 338.44
Formula Weight 338.2093
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.