missing translation for 'onlineSavingsMsg'
Learn More

Piperonyl Acetone, TRC

Product Code. 30426881
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30426881 2.5 g 2.5g
30426870 500 mg -
30418806 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30426881 Supplier Toronto Research Chemicals Supplier No. TRCP4753452.5G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55418-52-5
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.21
IUPAC Name 4-(1,3-benzodioxol-5-yl)butan-2-one
SMILES CC(=O)CCc1ccc2OCOc2c1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C11H12O3/c1-8(12)2-3-9-4-5-10-11(6-9)14-7-13-10/h4-6H,2-3,7H2,1H3
Formula Weight 192.0786
Chemical Name or Material Piperonyl Acetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.