missing translation for 'onlineSavingsMsg'
Learn More

Piperidolate, MedChemExpress

Product Code. 16468536
Encompass_Preferred
Click to view available options
Quantity:
100 mg
200 mg
Unit Size:
100mg
200mg
This item is not returnable. View return policy

Product Code. 16468536

Brand: Medchem Express HYB0962A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Piperidolate is an antimuscarinic, inhibits intestinal cramp induced by acetylcholine (rats and dogs).

Piperidolate is an antimuscarinic, inhibits intestinal cramp induced by acetylcholine (rats and dogs).

Specifications

Chemical Name or Material Piperidolate
Physical Form Viscous Liquid
CAS 82-98-4
Color Colorless
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C21H25NO2
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : ≥ 150 mg/mL (463.78 mM)
SMILES O=C(OC1CN(CC)CCC1)C(C2=CC=CC=C2)C3=CC=CC=C3
Molecular Weight (g/mol) 323.43
Formula Weight 323.43
Percent Purity 99.34%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.