missing translation for 'onlineSavingsMsg'
Learn More

Piperidino(4-piperidinyl)methanone, ≥97%, Thermo Scientific™

Product Code. 10041692
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041692 1 g Amber Glass Bottle 1g
10535733 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041692 Supplier Thermo Scientific Maybridge Supplier No. MO01170DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 63214-58-4
Molecular Formula C11H21N2O
Molecular Weight (g/mol) 197.30
MDL Number MFCD03372500
InChI Key JFHHGGAQABVJIF-UHFFFAOYSA-O
Synonym 1-piperidin-4-ylcarbonyl piperidine, piperidin-1-yl piperidin-4-yl methanone, piperidino 4-piperidinyl methanone, 1-piperidine-4-carbonyl piperidine, 4-piperidine-1-carbonyl piperidine, piperidin-4-yl-piperidin-1-yl-methanone, methanone,1-piperidinyl-4-piperidinyl, 1-piperidin-4-yl carbonyl piperidine, piperidine, 1-4-piperidinylcarbonyl
PubChem CID 2794688
SMILES O=C(C1CC[NH2+]CC1)N1CCCCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 4-(piperidine-1-carbonyl)piperidin-1-ium
Formula Weight 196.29
Percent Purity 97%
Chemical Name or Material Piperidino(4-piperidinyl)methanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.