missing translation for 'onlineSavingsMsg'
Learn More

(Piperidino-3-pyridinyl)methanol, 97%, Thermo Scientific™

Product Code. p-3075183
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10045652

Brand: Thermo Scientific Maybridge MO07116CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690631-99-3
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD05865147
InChI Key VLAMDMADWWGEDH-UHFFFAOYSA-N
Synonym 6-piperidin-1-yl-3-hydroxymethyl pyridine, 6-piperidin-1-yl pyridin-3-yl methanol, 6-piperidino-3-pyridinyl methanol, 6-piperidyl-3-pyridyl methan-1-ol, 3-pyridinemethanol,6-1-piperidinyl, 6-piperidin-1-ylpyridin-3-yl methanol
PubChem CID 2794792
IUPAC Name (6-piperidin-1-ylpyridin-3-yl)methanol
SMILES C1CCN(CC1)C2=NC=C(C=C2)CO

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material (Piperidino-3-pyridinyl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.