missing translation for 'onlineSavingsMsg'
Learn More

(Piperidino-3-pyridinyl)methanol, 97%, Thermo Scientific™

Product Code. 10265554
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10265554 10 g 10g
10045652 250 mg 250mg
10279742 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265554 Supplier Thermo Scientific Maybridge Supplier No. MO07116EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 690631-99-3
Molecular Formula C11H16N2O
Molecular Weight (g/mol) 192.262
MDL Number MFCD05865147
InChI Key VLAMDMADWWGEDH-UHFFFAOYSA-N
Synonym 6-piperidin-1-yl-3-hydroxymethyl pyridine, 6-piperidin-1-yl pyridin-3-yl methanol, 6-piperidino-3-pyridinyl methanol, 6-piperidyl-3-pyridyl methan-1-ol, 3-pyridinemethanol,6-1-piperidinyl, 6-piperidin-1-ylpyridin-3-yl methanol
PubChem CID 2794792
IUPAC Name (6-piperidin-1-ylpyridin-3-yl)methanol
SMILES C1CCN(CC1)C2=NC=C(C=C2)CO

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 10 g
Formula Weight 192.26
Percent Purity 97%
Chemical Name or Material (Piperidino-3-pyridinyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.