missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperidine, 99%, extra pure, Thermo Scientific Chemicals
301.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -11.0°C |
Density | 0.8620 g/mL |
Boiling Point | 106.0°C |
Flash Point | 16°C |
Assay Percent Range | 99% |
Packaging | Plastic drum |
Refractive Index | 1.4515 to 1.4535 |
Quantity | 10 L |
Beilstein | 20, 6 |
Fieser | 01,886; 02,332; 04,393; 06,472; 07,293 |
Show More |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'