missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Piperidine, 99%, extra pure
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
10 L
Unit Size:
1 L
10 L
100mL
2.5 L
500mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | -11.0°C |
| Density | 0.8620 g/mL |
| Boiling Point | 106.0°C |
| Flash Point | 16°C |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Refractive Index | 1.4515 to 1.4535 |
| Quantity | 1 L |
| Beilstein | 20, 6 |
| Fieser | 01,886; 02,332; 04,393; 06,472; 07,293 |
| Show More |
Safety and Handling
Product Identifier
- Piperidine
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P240-Ground/bond container and receiving equipment.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction