missing translation for 'onlineSavingsMsg'
Learn More

Piperidine, 99.5%, purified by redistillation, AcroSeal™

Product Code. 10411060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10411060 100 mL 100mL
10391280 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.
Product Code. 10411060 Supplier Thermo Scientific Acros Supplier No. 433841000
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains a category 2 drug precursor according to Regulation (EC) No 273/2004, all customers must complete a declaration before delivery.

CAS: 110-89-4 | C5H11N | 85.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-89-4
Molecular Formula C5H11N
Molecular Weight (g/mol) 85.15
InChI Key NQRYJNQNLNOLGT-UHFFFAOYSA-N
IUPAC Name piperidine
SMILES C1CCNCC1

Specifications

Melting Point -11.0°C
Density 0.8620 g/mL
Boiling Point 106.0°C
Flash Point 16°C
Assay Percent Range 99.5%
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4515 to 1.4535
Quantity 100 mL
Merck Index 15, 7580
Specific Gravity 0.862
Solubility Information Solubility aq. soln.: miscible. Other solubilities: soluble in alcohol and benzene, soluble in chloroform, acetone and ethanol
Viscosity 1.46 mPa.s (20°C)
Formula Weight 85.15
Chemical Name or Material Piperidine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Piperidine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.