missing translation for 'onlineSavingsMsg'
Learn More

Piperidine-1-sulfonyl chloride, 97%

Product Code. 15450478
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15450478 250 mg 250mg
15430478 1 g 1g
15440478 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15450478 Supplier Thermo Scientific Alfa Aesar Supplier No. H54053.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 35856-62-3 | C5H10ClNO2S | 183.65 g/mol

Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Piperidine-1-sulfonyl chloride is used as pharmaceutical intermediate.

Notes
Moisture sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35856-62-3
Molecular Formula C5H10ClNO2S
Molecular Weight (g/mol) 183.65
MDL Number MFCD03250329
InChI Key QQJYAXDCMMXECR-UHFFFAOYSA-N
Synonym 1-piperidinesulfonyl chloride, chloropiperidylsulfone, piperidinsulfochlorid, piperidinesulfonyl chloride, piperidinosulfonyl chloride, pperdne-1-sulfonyl chlorde, 1-piperidinesulfonylchloride, piperidinyl sulfonyl chloride, 1-piperidine sulfonyl chloride, piperidine-1-sulfonic acid chloride
PubChem CID 11298344
IUPAC Name piperidine-1-sulfonyl chloride
SMILES C1CCN(CC1)S(=O)(=O)Cl

Specifications

Density 1.308
Refractive Index 1.492
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 183.65
Percent Purity 97%
Chemical Name or Material Piperidine-1-sulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.