missing translation for 'onlineSavingsMsg'
Learn More

Piperazine-2-carboxylic acid dihydrochloride, 98%

Product Code. 10346780 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10346780 10 g 10g
10732875 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10346780 Supplier Thermo Scientific Acros Supplier No. 133660100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3022-15-9 | C5H10N2O2·2ClH | 203.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3022-15-9
Molecular Formula C5H10N2O2·2ClH
Molecular Weight (g/mol) 203.07
MDL Number MFCD00044803
InChI Key WNSDZBQLMGKPQS-UHFFFAOYSA-N
Synonym piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride
PubChem CID 2723757
IUPAC Name piperazine-2-carboxylic acid;dihydrochloride
SMILES C1CNC(CN1)C(=O)O.Cl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 200.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Glass bottle
Quantity 10 g
Formula Weight 203.07
Percent Purity 98%
Physical Form Crystalline Powder, Crystals or Chunks
Chemical Name or Material Piperazine-2-carboxylic acid dihydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Piperazine-2-carboxylic acid dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.