missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Piperazine-2-carboxylic acid dihydrochloride, 98%

Product Code. 11476180
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476180 25 g 25g
11466180 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476180 Supplier Thermo Scientific Alfa Aesar Supplier No. H27101.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3022-15-9 | C5H12Cl2N2O2 | 203.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3022-15-9
Molecular Formula C5H12Cl2N2O2
Molecular Weight (g/mol) 203.063
MDL Number MFCD00044803
InChI Key WNSDZBQLMGKPQS-UHFFFAOYSA-N
Synonym piperazine-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid dihydrochloride, piperazine-2-carboxylic acid 2hcl, +/--piperazine-2-carboxylic acid 2hcl, piperazin-2-carboxylic acid dihydrochloride, 2-piperazinecarboxylic acid, dihydrochloride, +/--2-piperazinecarboxylic acid dihydrochloride, +/--piperazine-2-carboxylic acid dihydrochloride, przca-2hcl, r-+-piperazine-2-carboxylic acid dihydrochloride
PubChem CID 2723757
IUPAC Name piperazine-2-carboxylic acid;dihydrochloride
SMILES C1CNC(CN1)C(=O)O.Cl.Cl

Specifications

Melting Point 270°C to 273°C
Quantity 25 g
Formula Weight 203.07
Percent Purity 98%
Chemical Name or Material (+/-)-Piperazine-2-carboxylic acid dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.