missing translation for 'onlineSavingsMsg'
Learn More

Pipemidic Acid Trihydrate, TRC

Product Code. 30134086
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134086 1 g 1g
17982785 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134086 Supplier Toronto Research Chemicals Supplier No. TRCP4798001G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 72571-82-5
Molecular Formula C14 H17 N5 O3 . 3 H2 O
Molecular Weight (g/mol) 357.36
Synonym Pipemidic Acid Trihydrate, 8-Ethyl-5,8-dihydro-5-oxo-2-(1-piperazinyl)pyrido[2,3-d]pyrimidine-6-carboxylic acid hydrate (1:3), 8-Ethyl-5,8-dihydro-5-oxo-2-(1-piperazinyl)pyrido[2,3-d]pyrimidine-6-carboxylic acid trihydrate, Pipemidic acid trihydrate
IUPAC Name 8-ethyl-5-oxo-2-piperazin-1-ylpyrido[2,3-d]pyrimidine-6-carboxylic acid;trihydrate
SMILES O.O.O.CCN1C=C(C(=O)O)C(=O)c2cnc(nc12)N3CCNCC3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H17N5O3.3H2O/c1-2-18-8-10(13(21)22)11(20)9-7-16-14(17-12(9)18)19-5-3-15-4-6-19;;;/h7-8,15H,2-6H2,1H3,(H,21,22);3*1H2
Formula Weight 357.1648
Chemical Name or Material Pipemidic acid trihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.