missing translation for 'onlineSavingsMsg'
Learn More

Pipemidic acid, 98%

Product Code. 15427799
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
This item is not returnable. View return policy

Product Code. 15427799

Brand: Thermo Scientific Alfa Aesar J66437.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51940-44-4 | C14H17N5O3 | 303.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51940-44-4
Molecular Formula C14H17N5O3
Molecular Weight (g/mol) 303.32
MDL Number MFCD00057291
InChI Key JOHZPMXAZQZXHR-UHFFFAOYSA-N
PubChem CID 4831
ChEBI CHEBI:75250
IUPAC Name 8-ethyl-5-oxo-2-(piperazin-1-yl)-5H,8H-pyrido[2,3-d]pyrimidine-6-carboxylic acid
SMILES CCN1C=C(C(O)=O)C(=O)C2=CN=C(N=C12)N1CCNCC1

Specifications

Type Pipemidic Acid
Physical Form Powder
Melting Point 251°C to 255°C
Packaging Plastic Bottle
Quantity 100 g
Flash Point >110°C (230°F)
Sensitivity Light sensitive
Boiling Point 717°C
Solubility Information Soluble in 1N sodium hydroxide or in DMSO. Insoluble in water
Formula Weight 303.32
Percent Purity 98%
Chemical Name or Material Pipemidic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.