missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Pipemarkers on a Card

Product Code. 15389085
Click to view available options
Display Text:
Argon
Bio gas
CO2
Carbon dioxide
Condensate
Cooling Gas
Freon
Freon 11
Gas
Helium
Inert Gas
LPG
Mixed Gas
Natural gas
Nitrogen
Recycled Gas
Return
Color:
Black on Ochre
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 15389085

Brand: Brady™ N007511

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

European Standard Pipemarkers reinforce safety and aid maintenance.

  • Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
  • Danger and hazard symbols , if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
  • Markers are printed with a contrasting border in the same color as the text for improved visibility.

Specifications

Certifications/Compliance ROHS Environmental Compliance, EU Directive 2002/95/EC, European Regulations 1271/2008, 790/2009 and 286/2001
Description Individual Pipe Markers on a Card with die-cut arrowheads, with pictograms - Gas - Gas
Adhesive Type Permanent Rubber, Permanent, Pressure-sensitive
Color Black on Ochre
Display Warning
Format Card
Material Polyester
Print Technology Thermal Transfer
Product Type Pipe Marker
Display Text Gas
For Use With (Application) Pipe and valve marking
Labels per Sheet 5
Languages English
No. per Pack 5 Markers/Card
Quantity 5 Pk.
Temperature Range (English) -40°F to +194°F
Temperature Range (Metric) -40°C to +90°C
Thickness (English) 0.0049 in.
Thickness (Metric) 0.124 mm
Dimensions (LxW) 12 x 150 mm
Width (English) 5.91 in.
Width (Metric) 150 mm
Height (English) 0.47 in.
Height (Metric) 12 mm
Height (Metric) Character 8 mm
Alternate Part Number GAS 150X12CARD-T1-P15
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.