missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ROBU VitraPOR™ Pipeline Filter
Pipeline Filter with tube adapters for connection.
Brand: ROBU VitraPOR™ 23604
This item is not returnable.
View return policy
Description
- Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Borosilicate Glass (3.3) | |
Tube Adapters | |
4 | |
60 mm |
Non-sterile | |
1 Ea. | |
230 mm |