missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pipecolic acid oxidase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99598-50ul
This item is not returnable.
View return policy
Description
Pipecolic acid oxidase Polyclonal antibody specifically detects Pipecolic acid oxidase in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Pipecolic acid oxidase | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| EC 1.5.3.1, EC 1.5.3.7, L-pipecolate oxidase, L-pipecolic acid oxidase, LPIPOXperoxisomal sarcosine oxidase, pipecolic acid oxidase, PSO | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Pipecolic acid oxidase. | |
| 50 μg | |
| Amino Acids Drugs and other small molecules, Cell Biology, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers | |
| 51268 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction