missing translation for 'onlineSavingsMsg'
Learn More

Pipecolic acid oxidase Antibody, Novus Biologicals™

Product Code. 18326689 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326689 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326689 Supplier Novus Biologicals Supplier No. H00051268D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Pipecolic acid oxidase Polyclonal antibody specifically detects Pipecolic acid oxidase in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Spécification

Antigen Pipecolic acid oxidase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot
Formulation PBS (pH 7.4)
Gene Accession No. NP_057602.2
Gene Alias EC 1.5.3.1, EC 1.5.3.7, L-pipecolate oxidase, L-pipecolic acid oxidase, LPIPOXperoxisomal sarcosine oxidase, pipecolic acid oxidase, PSO
Host Species Rabbit
Immunogen PIPOX (NP_057602.2, 1 a.a. - 390 a.a.) full-length human protein. MAAQKDLWDAIVIGAGIQGCFTAYHLAKHRKRILLLEQFFLPHSRGSSHGQSRIIRKAYLEDFYTRMMHECYQIWAQLEHEAGTQLHRQTGLLLLGMKENQELKTIQANLSRQRVEHQCLSSEELKQRFPNIRLPRGEVGLLDNSGGVIYAYKALRALQDAIRQLGGIVRDGEKVVEINPGLLVTVKTTSRSYQAKSLVITAGPWTNQLLRPLGIEMPLQTLRINVCYWREMVPGSYGVSQAFPCFLWLGLCPHHIYGLPTGEYPGLMKVSYHHGNHADPEERDCPTARTDIGDVQILSSFVRDHLPDLKPEPAVIESCMYTNTPDEQFILDRHPKYDNIVIGAGFSGHGFKLAPVVGKILYELSMKLTPSYDLAPFRISRFPSLGKAHL
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cell Biology, Core ESC Like Genes, Lipid and Metabolism, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 51268
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.