missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Pipe Markers (On Liner) - Standard Legends
Description
- Pipemarkers conform to RAL European standard color coding, and for dangerous substances, the Chemical Symbols conform to European Regulations 1271/2008, 790/2009 and 286/2001.
- Danger and hazard symbols, if relevant and necessary, are automatically printed either side of the text, and the Signal word underneath the text, which saves you having to identify the appropriate symbol.
- Markers are printed with a contrasting border in the same color as the text for improved visibility.
- Individual markers are supplied with a die-cut arrow head on both ends: simply remove the one not required
Specifications
Specifications
| Certifications/Compliance | ROHS Environmental Compliance, EU Directive 2002/95/EC, European Directives 67/548 and 88/379 |
| Description | Individual Pipe Markers on a Card with die-cut arrowheads, without pictograms - Acids and Alkalis - Acid |
| Adhesive Type | Permanent Acrylic, Permanent, Pressure-sensitive |
| Color | White on Purple |
| Finish | Gloss |
| Material | Polyester |
| Product Type | Pipe Marker |
| Display Text | Acid |
| For Use With (Application) | Pipe and valve marking |
| Labels per Sheet | 4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?