Learn More
SymLine™ Pipe Connection Pieces
Product Code. 18885890
Change view
Click to view available options
Closure Size:
3x (ID) 32 mm
Unit Size:
Each
Product Code. Closure Size unitSize
18885890 3x (ID) 32 mm Each
1 options
Product Code. 18885890 Supplier SymLine™ Supplier No. 106712
Only null left
Add to basket
Add to basket
These pipe connection pieces provide connection to the central collection point. Place the collecting container or canister where you want it: the pipe system can be exactly configured to suit your needs!
  • Wide range of connectors, branches and manifolds allow the pipe system to be connected precisely to the conditions in your laboratory
  • Specifications
    Closure Size 3x (ID) 32 mm
    Design T-piece
    Includes Sealing ring PTFE-EL and knurled nut
    Product Type Connection for Pipe System