missing translation for 'onlineSavingsMsg'
Learn More

Pipamperone, MedChemExpress

Product Code. 16485857
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16485857 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16485857 Supplier Medchem Express Supplier No. HY100703
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Pipamperone (Floropipamide; McN-JR 3345; R 3345) is a high-affinity antagonist of 5-HT2A receptor (pKi=8.2) and D4 receptor (pKi=8.0) and a low-affinity antagonist of D2 receptor (pKi=6.7).

Pipamperone (Floropipamide

Specifications

Chemical Name or Material Pipamperone
CAS 1893-33-0
Color White
Purity Grade Notes Research
Molecular Formula C21H30FN3O2
Quantity 5 mg
Synonym Floropipamide McN-JR 3345 R 3345
Solubility Information DMSO : 100 mg/mL (266.33 mM; Need ultrasonic) ∣0.1 M HCL : 25 mg/mL (66.58 mM; Need ultrasonic)
SMILES O=C(C1(N2CCCCC2)CCN(CCCC(C3=CC=C(F)C=C3)=O)CC1)N
Molecular Weight (g/mol) 375.48
Formula Weight 375.48
Percent Purity 98.96%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.